missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 4-Pentylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09810DA

 View more versions of this product

Catalog No. BTB09810DA



Description

Description

Specifications

Specifications

4-Pentylbenzene-1-sulfonyl chloride
96.5
97%
C11H15ClO2S
MFCD00173759
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
246.75
246.76
73948-18-2
100.0
Amber glass bottle
CH3(CH2)4C6H4SO2Cl
1 g
XDWXZRVWJHEWMT-UHFFFAOYSA-N
4-pentylbenzenesulfonyl chloride
144625
97%

Chemical Identifiers

73948-18-2
246.75
XDWXZRVWJHEWMT-UHFFFAOYSA-N
144625
C11H15ClO2S
MFCD00173759
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions