Please login to your online account to display your discounted pricing

4-Pentylbenzene-1-sulfonyl chloride, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB09810EA

 View more versions of this product

Catalog No. BTB09810EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 246.76
Quantity 10g
Assay 97%
CAS Max % 100.0
Linear Formula CH3(CH2)4C6H4SO2Cl
Chemical Name or Material 4-Pentylbenzene-1-sulfonyl chloride
Assay Percent Range 97%
MDL Number MFCD00173759
CAS Min % 96.5
Packaging Amber glass bottle
Molecular Formula C11H15ClO2S
CAS 73948-18-2
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Pentylbenzene-1-sulfonyl chloride
CAS 73948-18-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H15ClO2S
Linear Formula CH3(CH2)4C6H4SO2Cl
Formula Weight 246.76
MDL Number MFCD00173759
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents