Learn More
4-Pentylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
$15.04 - $57.86
Chemical Identifiers
CAS | 73948-18-2 |
---|---|
Molecular Formula | C11H15ClO2S |
Molecular Weight (g/mol) | 246.75 |
MDL Number | MFCD00173759 |
InChI Key | XDWXZRVWJHEWMT-UHFFFAOYSA-N |
Synonym | 4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride |
PubChem CID | 144625 |
SMILES | CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BTB09810DA
|
Thermo Scientific Chemicals
BTB09810DA |
1 g |
Each for $15.04
|
|
BTB09810EA
|
Thermo Scientific Chemicals
BTB09810EA |
10 g |
Each for $57.86
|
|
Chemical Identifiers
73948-18-2 | |
246.75 | |
XDWXZRVWJHEWMT-UHFFFAOYSA-N | |
144625 |
C11H15ClO2S | |
MFCD00173759 | |
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride | |
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
Specifications
73948-18-2 | |
100.0 | |
C11H15ClO2S | |
MFCD00173759 | |
1 g | |
XDWXZRVWJHEWMT-UHFFFAOYSA-N | |
4-pentylbenzenesulfonyl chloride | |
144625 | |
97% |
96.5 | |
97% | |
CH3(CH2)4C6H4SO2Cl | |
Amber glass bottle | |
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride | |
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O | |
246.75 | |
246.76 | |
4-Pentylbenzene-1-sulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.