missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Pentenyl)triphenylphosphonium bromide, 97%

$78.96 - $1,084.11

Chemical Identifiers

CAS 56771-29-0
Molecular Formula C23H24BrP
Molecular Weight (g/mol) 411.323
MDL Number MFCD00050215
InChI Key KANICXDUFCDRDS-UHFFFAOYSA-M
Synonym 4-pentenyl triphenylphosphonium bromide, pent-4-enyl triphenyl phosphonium bromide, pent-4-en-1-yltriphenylphosphonium bromide, pent-4-en-1-yltriphenylphosphanium bromide, acmc-1az5e, pent-4-enyl triphenyl phosphanium bromide, 4-?pentenyl ?triphenylphosphonium bromide, 4-pentenyl triphenylphosphonium bromide, phosphonium,4-penten-1-yltriphenyl-, bromide 1:1
PubChem CID 10093022
IUPAC Name pent-4-enyl(triphenyl)phosphanium;bromide
SMILES C=CCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0195003 View Documents Alfa Aesar™
L0195003
1g Each for $78.96
 
AAL0195006 View Documents Alfa Aesar™
L0195006
5g Each for $263.70
 
AAL0195014 View Documents Alfa Aesar™
L0195014
25g Each for $1,084.11
 
Specifications

Chemical Identifiers

56771-29-0
411.323
KANICXDUFCDRDS-UHFFFAOYSA-M
10093022
C=CCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C23H24BrP
MFCD00050215
4-pentenyl triphenylphosphonium bromide, pent-4-enyl triphenyl phosphonium bromide, pent-4-en-1-yltriphenylphosphonium bromide, pent-4-en-1-yltriphenylphosphanium bromide, acmc-1az5e, pent-4-enyl triphenyl phosphanium bromide, 4-?pentenyl ?triphenylphosphonium bromide, 4-pentenyl triphenylphosphonium bromide, phosphonium,4-penten-1-yltriphenyl-, bromide 1:1
pent-4-enyl(triphenyl)phosphanium;bromide

Specifications

194°C to 196°C
56771-29-0
MFCD00050215
4-pentenyl triphenylphosphonium bromide, pent-4-enyl triphenyl phosphonium bromide, pent-4-en-1-yltriphenylphosphonium bromide, pent-4-en-1-yltriphenylphosphanium bromide, acmc-1az5e, pent-4-enyl triphenyl phosphanium bromide, 4-?pentenyl ?triphenylphosphonium bromide, 4-pentenyl triphenylphosphonium bromide, phosphonium,4-penten-1-yltriphenyl-, bromide 1:1
C=CCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
411.323
411.33
(4-Pentenyl)triphenylphosphonium bromide
1g
C23H24BrP
4171464
KANICXDUFCDRDS-UHFFFAOYSA-M
pent-4-enyl(triphenyl)phosphanium;bromide
10093022
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning