missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Pentenoic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC269021000
Specifications
4-Pentenoic acid | |
89°C | |
Glass bottle | |
0.975 | |
Colorless to Yellow | |
100g | |
98.5% min. (GC) | |
H2C=CHCH2CH2CO2H | |
.delta.4-pentenoic acid, 3-vinylpropionic acid, 4 pa, 4-pentenoic acid, allyl acetic acid, allylacetic acid, delta 4-pentenoic acid, fema no. 2843, unii-d4s77y29fb, wln: qv3u1 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
pent-4-enoic acid | |
61138 | |
100.2 | |
99% |
0.9750g/mL | |
Authentic | |
1.4273 to 1.4303 | |
83.0°C to 84.0°C (12.0mmHg) | |
-22.5°C | |
591-80-0 | |
C5H8O2 | |
02,425 | |
Solubility in water: insoluble | |
C=CCCC(=O)O | |
100.2 | |
CHEBI:35936 | |
Liquid |
Chemical Identifiers
591-80-0 | |
100.2 | |
.delta.4-pentenoic acid, 3-vinylpropionic acid, 4 pa, 4-pentenoic acid, allyl acetic acid, allylacetic acid, delta 4-pentenoic acid, fema no. 2843, unii-d4s77y29fb, wln: qv3u1 | |
CHEBI:35936 | |
C=CCCC(=O)O |
C5H8O2 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
61138 | |
pent-4-enoic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 209-732-7