missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Penten-2-ol, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 387430250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Penten-2-ol | |
95.0 | |
0.8370g/mL | |
25°C | |
95% min. (GC) | |
C5H10O | |
H2C=CHCH2CH(OH)CH3 | |
25g | |
4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa | |
ZHZCYWWNFQUZOR-UHFFFAOYSA-N | |
pent-4-en-2-ol | |
12247 | |
96% |
625-31-0 | |
100.0 | |
115.0°C to 116.0°C | |
Authentic | |
Glass bottle | |
1.4230 to 1.4250 | |
MFCD00004556 | |
0.837 | |
Solubility in water: fully miscible | |
CC(CC=C)O | |
86.13 | |
86.13 |
Chemical Identifiers
625-31-0 | |
86.13 | |
ZHZCYWWNFQUZOR-UHFFFAOYSA-N | |
12247 | |
CC(CC=C)O |
C5H10O | |
MFCD00004556 | |
4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa | |
pent-4-en-2-ol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Store in a well-ventilated place.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 210-887-8
RUO â Research Use Only