missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Penten-1-yl Isothiocyanate, TCI America™

$352.92 - $1,084.75

Chemical Identifiers

CAS 18060-79-2
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00191527
InChI Key DBISBKDNOKIADM-UHFFFAOYSA-N
Synonym Isothiocyanic Acid 4-Penten-1-yl Ester
PubChem CID 87436
IUPAC Name 5-isothiocyanatopent-1-ene
SMILES C=CCCCN=C=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04441G View Documents TCI America
I04441G
1g Each for $352.92
 
I04445G View Documents TCI America
I04445G
5g Each for $1,084.75
 
Specifications

Chemical Identifiers

18060-79-2
127.205
DBISBKDNOKIADM-UHFFFAOYSA-N
87436
C=CCCCN=C=S
C6H9NS
MFCD00191527
Isothiocyanic Acid 4-Penten-1-yl Ester
5-isothiocyanatopent-1-ene

Specifications

194°C
1g
C6H9NS
1992
DBISBKDNOKIADM-UHFFFAOYSA-N
5-isothiocyanatopent-1-ene
87436
Clear Liquid at 20°C
4-Penten-1-yl Isothiocyanate
Yellow
18060-79-2
MFCD00191527
Isothiocyanic Acid 4-Penten-1-yl Ester
C=CCCCN=C=S
127.205
127.21
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No