Please login to your online account to display your discounted pricing

4-Penten-1-yl Acetate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Penten-1-yl acetate
Name Note 98%
CAS 1576-85-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H12O2
Linear Formula CH3CO2(CH2)3CH=CH2
Formula Weight 128.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149720100

  SDS
thermo fisher scientific
149720100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 128.17
Quantity 10g
Color Clear yellow
Beilstein 02, II, 151
Density 0.91g/mL
EINECS Number 216-413-6
CAS Max % 100.0
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
Name Note 98%
Molecular Formula C7H12O2
Linear Formula CH3CO2(CH2)3CH=CH2
MDL Number MFCD00008723
CAS 1576-85-8
Synonym 5-Acetoxy-1-pentene
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 4-Penten-1-yl acetate
Assay Percent Range 98%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.