missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Penten-1-ol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150010500

 View more versions of this product

Catalog No. AC150010500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Penten-1-ol
821-09-0
100.0
0.8340g/mL
43°C
98.5% min. (GC)
C5H10O
H2C=CH(CH2)3OH
50 g
0.834
Solubility in water: 57g/L (25°C)
C=CCCCO
86.13
86.13
Liquid
99%
98.5
Colorless
134°C to 137°C
Authentic
Glass bottle
1.4289 to 1.4309
MFCD00002975
01, 443
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
LQAVWYMTUMSFBE-UHFFFAOYSA-N
pent-4-en-1-ol
13181
99%

Chemical Identifiers

821-09-0
86.13
LQAVWYMTUMSFBE-UHFFFAOYSA-N
13181
C=CCCCO
C5H10O
MFCD00002975
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
pent-4-en-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 212-473-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only