Please login to your online account to display your discounted pricing

4-P-PDOT, Tocris


MT2 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS103410

 View more versions of this product

Catalog No. 103410

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 279.38
Quantity 10mg
Chemical Name or Material 4-P-PDOT
Assay Percent Range >99%
Molecular Formula C19H21NO
CAS 134865-74-0
Synonym cis-4-Phenyl-2-propionamidotetralin
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 4-P-PDOT
Assay Percent Range >99%
Molecular Formula C19H21NO
Formula Weight 279.38
Synonym cis-4-Phenyl-2-propionamidotetralin
Accessories

Store at Room Temperature