Please login to your online account to display your discounted pricing

4-P-PDOT, Tocris

MT2 antagonist

$175.98 - $750.85

Chemical Identifiers

Chemical Name or Material 4-P-PDOT
Assay Percent Range >99%
Molecular Formula C19H21NO
Formula Weight 279.38
Synonym cis-4-Phenyl-2-propionamidotetralin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

103410

 
r & d systems
RDS103410
10mg Each for $175.98

103450

 
r & d systems
RDS103450
50mg Each for $750.85
Description & Specifications

Specifications

Formula Weight 279.38
Quantity 10mg
Chemical Name or Material 4-P-PDOT
Assay Percent Range >99%
Molecular Formula C19H21NO
CAS 134865-74-0
Synonym cis-4-Phenyl-2-propionamidotetralin
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature