Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Oxocyclohexanecarboxylic acid, 98%

$85.74 - $215.50

Chemical Identifiers

Chemical Name or Material 4-Oxocyclohexanecarboxylic acid
CAS 874-61-3
Molecular Formula C7H10O3
Formula Weight 142.16
MDL Number MFCD00102035
Synonym Cyclohexanone-4-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27294MD

  SDS
alfa aesar™
H27294MD
250mg Each for $85.74

AAH2729403

  SDS
alfa aesar™
H2729403
1g Each for $215.50
Description & Specifications

Specifications

Formula Weight 142.16
Chemical Name or Material 4-Oxocyclohexanecarboxylic acid
Assay 98%
Molecular Formula C7H10O3
MDL Number MFCD00102035
CAS 874-61-3
Synonym Cyclohexanone-4-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 65° to 71°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.