Please login to your online account to display your discounted pricing

4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW02412CB

 View more versions of this product

Catalog No. SEW02412CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.16
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C9H8O4
MDL Number MFCD00052170
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid
Assay Percent Range 97%
CAS 56671-28-4
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid
CAS 56671-28-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8O4
Formula Weight 180.16
MDL Number MFCD00052170
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents