Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Oxo-4-(2-thienyl)butyric acid, 98%

$92.97 - $343.54

Chemical Identifiers

Chemical Name or Material 4-Oxo-4-(2-thienyl)butyric acid
CAS 4653-08-1
Molecular Formula C8H8O3S
Formula Weight 184.21
MDL Number MFCD00022502
Beilstein 133870
Synonym 3-(2-Thenoyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0310014

  SDS
alfa aesar™
L0310014
25g Each for $343.54

AAL0310006

  SDS
alfa aesar™
L0310006
5g Each for $92.97
Description & Specifications

Specifications

Formula Weight 184.21
Chemical Name or Material 4-Oxo-4-(2-thienyl)butyric acid
CAS 4653-08-1
Molecular Formula C8H8O3S
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym 3-(2-Thenoyl)propionic acid
Beilstein 133870
Assay 98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Odorless
Quantity 25g
MDL Number MFCD00022502
Melting Point 116° to 121°C
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.