missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268110010

 View more versions of this product

Catalog No. AC268110010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene
Glass bottle
117.0°C to 122.0°C
99%
99%
MFCD00143363
Solubility in water: .. Other solubilities: soluble in benzene and ethanol
C1C2C=CC1C3C2C(=O)N3
130862428
Needle-Like Crystalline Powder
Authentic
Beige
1g
14735-70-7
C8H9NO
3-aza-tricyclo 4.2.1.0 2,5 non-7-en-4-one
WBZQHVKGKQXWMW-IBJXHFRJSA-N
135.17
135.17
99%

Chemical Identifiers

14735-70-7
135.17
WBZQHVKGKQXWMW-IBJXHFRJSA-N
130862428
C8H9NO
MFCD00143363
3-aza-tricyclo 4.2.1.0 2,5 non-7-en-4-one
C1C2C=CC1C3C2C(=O)N3
SDS Documents