missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 99%
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00143363
Chemical Name or Material 4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC268110010 SDS Fisher Scientific Acros
268110010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC268110050 SDS Thermo Fisher Scientific
AC268110050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

99%
135.17
4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%
C8H9NO
MFCD00143363
≥98.5%

Specifications

Needle-Like Crystalline Powder
1g
117° to 122°C
C8H9NO
MFCD00143363
≥98.5%
135.17
Slightly beige
Authentic
99%
4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%