Please login to your online account to display your discounted pricing

4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%, free radical, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  176860050

 View more versions of this product

Catalog No. AC176860050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Needles, Crystals or Crystalline Powder
Formula Weight 170.23
Quantity 5g
Color Orange
TSCA TSCA
EINECS Number 220-778-7
RTECS Number TN9200000
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Synonym 2,2,6,6-Tetramethyl-4-oxopiperidinooxy
Infrared Spectrum Authentic
Molecular Formula C9H16NO2
CAS 2896-70-0
Chemical Name or Material 4-Oxo-2,2,6,6-tetramethylpiperidinooxy
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00006187

Specifications

Chemical Name or Material 4-Oxo-2,2,6,6-tetramethylpiperidinooxy
Name Note 95%
CAS 2896-70-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H16NO2
Formula Weight 170.23
MDL Number MFCD00006187
Synonym 2,2,6,6-Tetramethyl-4-oxopiperidinooxy