missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%, free radical, ACROS Organics™

Manufacturer:  Acros Organics 176860010

 View more versions of this product

Catalog No. AC176860010


 

Description

Description

Polymerization reagent
Specifications

Specifications

4-Oxo-2,2,6,6-tetramethylpiperidinooxy
2896-70-0
100.0
≥94%
C9H16NO2
Glass bottle
170.23
Orange
95%
94.0
Authentic
95%
MFCD00006187
2,2,6,6-Tetramethyl-4-oxopiperidinooxy, 4-Oxo-TEMPO
Needles, Crystals or Crystalline Powder
1g

Chemical Identifiers

2896-70-0
C9H16NO2
MFCD00006187
2,2,6,6-Tetramethyl-4-oxopiperidinooxy, 4-Oxo-TEMPO
Safety and Handling

Safety and Handling

EINECSNumber : 220-778-7

RTECSNumber : TN9200000

TSCA : TSCA

SDS Documents