Please login to your online account to display your discounted pricing

4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%, free radical, ACROS Organics™

$67.29 - $201.03

Chemical Identifiers

Chemical Name or Material 4-Oxo-2,2,6,6-tetramethylpiperidinooxy
Name Note 95%
CAS 2896-70-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H16NO2
Formula Weight 170.23
MDL Number MFCD00006187
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176860010

  SDS
thermo fisher scientific
176860010
Glass bottle 1g Each for $67.29

AC176860050

  SDS
thermo fisher scientific
176860050
Glass bottle 5g Each for $201.03
Description & Specifications

Specifications

Physical Form Needles, Crystals or Crystalline Powder
Assay ≥94%
Formula Weight 170.23
Quantity 1g
Color Orange
EINECS Number 220-778-7
TSCA TSCA
RTECS Number TN9200000
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 4-Oxo-2,2,6,6-tetramethylpiperidinooxy
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H16NO2
MDL Number MFCD00006187
CAS 2896-70-0
Packaging Glass bottle
Synonym 2,2,6,6-Tetramethyl-4-oxopiperidinooxy

Polymerization reagent