missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 95.0+%, TCI America™

Manufacturer:  TCI America O026625G

 View more versions of this product

Catalog No. O026625G



Specifications

Specifications

4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
37°C
2896-70-0
MFCD00006187
4-Oxo-TEMPO
Crystal-Powder at 20°C
Undesignated
25g
C9H16NO2
3077
170.23
≥95.0% (GC,T)

Chemical Identifiers

2896-70-0
C9H16NO2
MFCD00006187
4-Oxo-TEMPO
Safety and Handling

Safety and Handling

RTECSNumber : TN9200000

TSCA : Yes

Recommended Storage : Freezer

Documents