missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxo-1-phenylcyclohexanecarbonitrile 98.0+%, TCI America™

Manufacturer:  TCI America O04705G

 View more versions of this product

Catalog No. O04705G



Specifications

Specifications

4-Oxo-1-phenylcyclohexanecarbonitrile
119°C
25115-74-6
MFCD00044816
GKXOABVSZWCJJK-UHFFFAOYSA-N
4-oxo-1-phenylcyclohexane-1-carbonitrile
91282
Crystal-Powder at 20°C
White
5g
C13H13NO
4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone
C1CC(CCC1=O)(C#N)C2=CC=CC=C2
199.253
199.25
≥98.0% (GC)

Chemical Identifiers

25115-74-6
199.253
GKXOABVSZWCJJK-UHFFFAOYSA-N
91282
C1CC(CCC1=O)(C#N)C2=CC=CC=C2
C13H13NO
MFCD00044816
4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone
4-oxo-1-phenylcyclohexane-1-carbonitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents