missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Oxazolecarboxaldehyde, TCI America™

Manufacturer:  TCI America O04531G

 View more versions of this product

Catalog No. O04531G



Specifications

Specifications

4-Oxazolecarboxaldehyde
60°C
118994-84-6
MFCD02179512
JBWIIXBEPINWPB-UHFFFAOYSA-N
1,3-oxazole-4-carbaldehyde
14281432
Crystal-Powder at 20°C
Yellow
1g
C4H3NO2
4-Formyloxazole
C1=C(N=CO1)C=O
97.073
97.07
≥98.0% (GC)

Chemical Identifiers

118994-84-6
97.073
JBWIIXBEPINWPB-UHFFFAOYSA-N
14281432
C1=C(N=CO1)C=O
C4H3NO2
MFCD02179512
4-Formyloxazole
1,3-oxazole-4-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents