Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Oxatetradecanoic acid, 99%

Chemical Identifiers

Chemical Name or Material 4-Oxatetradecanoic acid
CAS 7420-16-8
Formula Weight 230.35
MDL Number MFCD00214355
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67471MA

 
alfa aesar™
J67471MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.35
Chemical Name or Material 4-Oxatetradecanoic acid
CAS 7420-16-8
MDL Number MFCD00214355
Assay 99%
Physical Form Solid
Storage Note 1 -30° to -10°C
Quantity 10mg
Safety and Handling

-30° to -10°C