missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Octyne, TCI America™

Manufacturer:  TCI America O012725ML

 View more versions of this product

Catalog No. O012725ML



Specifications

Specifications

4-Octyne
Colorless
25mL
C8H14
3295
GZTNBKQTTZSQNS-UHFFFAOYSA-N
oct-4-yne
16029
Clear Liquid at 20°C
131°C
-103°C
1942-45-6
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, 4-octyne, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
CCCC#CCCC
110.2
110.20
≥99.0% (GC)

Chemical Identifiers

1942-45-6
110.2
GZTNBKQTTZSQNS-UHFFFAOYSA-N
16029
CCCC#CCCC
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, 4-octyne, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
oct-4-yne
Safety and Handling

Safety and Handling

TSCA : Yes

Documents