Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Octen-4-ylboronic acid pinacol ester, 98%

$110.14 - $293.82

Chemical Identifiers

Chemical Name or Material 4-Octen-4-ylboronic acid pinacol ester
CAS 177949-95-0
Molecular Formula C14H27BO2
Formula Weight 238.18
MDL Number MFCD03788750
Beilstein 9128681
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1969903

  SDS
alfa aesar™
L1969903
1g Each for $293.82

AAL19699MD

  SDS
alfa aesar™
L19699MD
250mg Each for $110.14
Description & Specifications

Specifications

Formula Weight 238.18
MDL Number MFCD03788750
CAS 177949-95-0
Assay 98%
Molecular Formula C14H27BO2
Quantity 1g
Beilstein 9128681
Chemical Name or Material 4-Octen-4-ylboronic acid pinacol ester