Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Octen-4-ylboronic acid diethanolamine ester, 97%

$79.24 - $79.24

Chemical Identifiers

Chemical Name or Material 4-Octen-4-ylboronic acid diethanolamine ester
CAS 608534-40-3
Molecular Formula C12H24BNO2
Formula Weight 225.14
MDL Number MFCD03788756
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19705MC

 
alfa aesar™
L19705MC
100mg Each for $79.24
Description & Specifications

Specifications

Formula Weight 225.14
Chemical Name or Material 4-Octen-4-ylboronic acid diethanolamine ester
Molecular Formula C12H24BNO2
MDL Number MFCD03788756
Assay 97%
CAS 608534-40-3
Melting Point 160° to 167°C
Quantity 100mg