Please login to your online account to display your discounted pricing

4-Octanol, +98%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC389620100

 View more versions of this product

Catalog No. AC389620100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Liquid
Formula Weight 130.23
Quantity 10mL
Color Clear colorless
Density 0.8180g/mL
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 4-Octanol
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98+%
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
MDL Number MFCD00014409
CAS 589-62-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Octanol
CAS 589-62-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Formula Weight 130.23
MDL Number MFCD00014409
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents