Please login to your online account to display your discounted pricing

4-Octanol, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Octanol
CAS 589-62-8
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Formula Weight 130.23g/mol
MDL Number MFCD00014409
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389620100

  SDS
thermo fisher scientific
389620100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.8180g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Color Clear colorless
Quantity 10mL
Chemical Name or Material 4-Octanol
CAS 589-62-8
CAS Min % 98.0
CAS Max % 100.0
Physical Form Clear colorless liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Formula Weight 130.23g/mol
MDL Number MFCD00014409
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.