missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Octadecyloxybiphenyl, TCI America™

$105.42 - $323.89

Chemical Identifiers

CAS 376609-78-8
Molecular Formula C30H46O
Molecular Weight (g/mol) 422.697
MDL Number MFCD01321145
InChI Key WJPFTOGDAJEWPP-UHFFFAOYSA-N
Synonym Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether, 4-Stearyloxybiphenyl
PubChem CID 22899691
IUPAC Name 1-octadecoxy-4-phenylbenzene
SMILES CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19245G View Documents TCI America
B19245G
5g Each for $105.42
 
B192425G View Documents TCI America
B192425G
25g Each for $323.89
 
Specifications

Chemical Identifiers

376609-78-8
422.697
WJPFTOGDAJEWPP-UHFFFAOYSA-N
22899691
CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C2=CC=CC=C2
C30H46O
MFCD01321145
Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether, 4-Stearyloxybiphenyl
1-octadecoxy-4-phenylbenzene

Specifications

White
5g
C30H46O
Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether, 4-Stearyloxybiphenyl
CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C2=CC=CC=C2
422.697
422.70
≥98.0% (GC)
92°C
376609-78-8
MFCD01321145
WJPFTOGDAJEWPP-UHFFFAOYSA-N
1-octadecoxy-4-phenylbenzene
22899691
Crystal-Powder at 20°C
4-Octadecyloxybiphenyl
Safety and Handling

Safety and Handling

TSCA : No