missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-O-Benzyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal, 97%

$57.97 - $183.51

Chemical Identifiers

CAS 111830-58-1
Molecular Formula C25H44O4Si2
Molecular Weight (g/mol) 464.793
MDL Number MFCD22989001
InChI Key CZGTZYNJABAJPH-ZLNRFVROSA-N
Synonym 4-o-benzyl-3,6-di-o-tert-butyldimethylsilyl-d-glucal, 3-o,6-o-bis tert-butyldimethylsilyl-4-o-benzyl-1,2-dideoxy-1,2-didehydro-d-glucopyranose, 2r,3r,4r-3-benzyloxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-4-yl oxy tert-butyl dimethylsilane
PubChem CID 11113360
IUPAC Name tert-butyl-[[(2R,3R,4R)-2-[[tert-butyl(dimethyl)silyl]oxymethyl]-3-phenylmethoxy-3,4-dihydro-2H-pyran-4-yl]oxy]-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60280MD View Documents Alfa Aesar™
H60280MD
250mg Each for $57.97
 
AAH6028003 View Documents Alfa Aesar™
H6028003
1g Each for $183.51
 
Specifications

Chemical Identifiers

111830-58-1
464.793
CZGTZYNJABAJPH-ZLNRFVROSA-N
11113360
CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)OCC2=CC=CC=C2
C25H44O4Si2
MFCD22989001
4-o-benzyl-3,6-di-o-tert-butyldimethylsilyl-d-glucal, 3-o,6-o-bis tert-butyldimethylsilyl-4-o-benzyl-1,2-dideoxy-1,2-didehydro-d-glucopyranose, 2r,3r,4r-3-benzyloxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-4-yl oxy tert-butyl dimethylsilane
tert-butyl-[[(2R,3R,4R)-2-[[tert-butyl(dimethyl)silyl]oxymethyl]-3-phenylmethoxy-3,4-dihydro-2H-pyran-4-yl]oxy]-dimethylsilane

Specifications

250mg
C25H44O4Si2
4-o-benzyl-3,6-di-o-tert-butyldimethylsilyl-d-glucal, 3-o,6-o-bis tert-butyldimethylsilyl-4-o-benzyl-1,2-dideoxy-1,2-didehydro-d-glucopyranose, 2r,3r,4r-3-benzyloxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-4-yl oxy tert-butyl dimethylsilane
CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)OCC2=CC=CC=C2
464.793
464.79
4-O-Benzyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal
111830-58-1
MFCD22989001
CZGTZYNJABAJPH-ZLNRFVROSA-N
tert-butyl-[[(2R,3R,4R)-2-[[tert-butyl(dimethyl)silyl]oxymethyl]-3-phenylmethoxy-3,4-dihydro-2H-pyran-4-yl]oxy]-dimethylsilane
11113360
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C