missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-O-Acetyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal, 97%

$55.51 - $175.80

Chemical Identifiers

CAS 132891-79-3
Molecular Formula C20H40O5Si2
MDL Number MFCD01321282
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60975MD View Documents Alfa Aesar™
H60975MD
250mg Each for $55.51
 
AAH6097503 View Documents Alfa Aesar™
H6097503
1g Each for $175.80
 
Specifications

Chemical Identifiers

132891-79-3
C20H40O5Si2
MFCD01321282

Specifications

>113°C (235°F)
324°C
132891-79-3
MFCD01321282
97%
−19° (c=2.1 in Chloroform)
250mg
C20H40O5Si2
416.7
4-O-Acetyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C