missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-O-Acetyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal, 97%

Manufacturer:  Alfa Aesar™ H6097503

 View more versions of this product

Catalog No. AAH6097503



Specifications

Specifications

4-O-Acetyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal
97%
MFCD01321282
324°C
>113°C (235°F)
132891-79-3
C20H40O5Si2
416.7
1g
−19° (c=2.1 in Chloroform)

Chemical Identifiers

132891-79-3
C20H40O5Si2
MFCD01321282
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

SDS Documents