Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nonyne, 96%

Chemical Identifiers

Chemical Name or Material 4-Nonyne
CAS 20184-91-2
Molecular Formula C9H16
Linear Formula CH3(CH2)3C≡CCH2CH2CH3
Formula Weight 124.23g/mol
MDL Number MFCD00041650
UN Number UN3295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2134906

 
alfa aesar™
B21349-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 158°C
Quantity 5g
Flash Point 35°C (95°F)
Chemical Name or Material 4-Nonyne
CAS 20184-91-2
Assay 96%
Molecular Formula C9H16
Linear Formula CH3(CH2)3C≡CCH2CH2CH3
Formula Weight 124.23g/mol
MDL Number MFCD00041650
UN Number UN3295
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.