Please login to your online account to display your discounted pricing

4-Nonylphenol, Mixture of isomers, 99%, ACROS Organics™


Quantity 5kg
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC416240050

 View more versions of this product

Catalog No. AC416240050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 220.35
Physical Form Viscous Liquid
Quantity 5kg
Color Clear
Packaging Plastic Jerrycan
Density 0.9400g/mL
Merck Index 14, 6681
CAS Max % 100.0
Water 0.05% max.
Refractive Index 1.5105 to 1.5165 (20°C, 589nm)
CAS 84852-15-3
Chemical Name or Material 4-Nonylphenol
MDL Number MFCD00002396
Name Note Mixture of isomers
CAS Min % 98.5
Molecular Formula C15H24O
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility. Suspected of damaging the unborn child.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Nonylphenol
Name Note Mixture of isomers
CAS 84852-15-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C15H24O
Formula Weight 220.35
MDL Number MFCD00002396
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility. Suspected of damaging the unborn child.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents