Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nonanone, 97+%

$62.68 - $611.74

Chemical Identifiers

Chemical Name or Material 4-Nonanone
CAS 4485-09-0
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00027286
Synonym n-Pentyl n-propyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1505014

  SDS
alfa aesar™
A1505014
25g Each for $62.68

AAA1505022

  SDS
alfa aesar™
A1505022
100g Each for $153.69

AAA1505036

  SDS
alfa aesar™
A1505036
500g Each for $611.74
Description & Specifications

Specifications

Formula Weight 142.24
MDL Number MFCD00027286
Chemical Name or Material 4-Nonanone
Molecular Formula C9H18O
Synonym n-Pentyl n-propyl ketone
TSCA TSCA
Boiling Point 187°C
Assay ≥97%
Quantity 25g
CAS 4485-09-0