Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nonanol, 95%

$223.19 - $935.88

Chemical Identifiers

Chemical Name or Material 4-Nonanol
CAS 5932-79-6
Molecular Formula C9H20O
Formula Weight 144.26
MDL Number MFCD00021950
Beilstein 1719438
Synonym n-Amyl n-propyl carbinol; n-Pentyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1949618

 
alfa aesar™
A1949618
50g Each for $223.19

AAA1949630

 
alfa aesar™
A1949630
250g Each for $935.88
Description & Specifications

Specifications

Formula Weight 144.26
Chemical Name or Material 4-Nonanol
TSCA TSCA
Assay 95%
Molecular Formula C9H20O
MDL Number MFCD00021950
CAS 5932-79-6
Beilstein 1719438
Synonym n-Amyl n-propyl carbinol; n-Pentyl n-propyl carbinol
Flash Point 74°C (165°F)
Boiling Point 192° to 195°C
Quantity 50g