Please login to your online account to display your discounted pricing

4-Nitrothiophenol, 80%, Tech., ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  336200250

 View more versions of this product

Catalog No. AC336200250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥75%
Formula Weight 155.2
Physical Form Crystalline Powder and/or Chunks
Purity Grade Technical Grade
Quantity 25g
Color Yellow
Beilstein 06, 339
EINECS Number 217-436-4
Melting Point 72° to 80°C
RTECS Number DC1850000
CAS Max % 100.0
CAS Min % 75.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 4-Nitrobenzenethiol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 STENCH
Linear Formula SHC6H4NO
Molecular Formula C6H5NO2S
Chemical Name or Material 4-Nitrothiophenol
Name Note 95%
CAS 1849-36-1
Assay Percent Range 80%
MDL Number MFCD00007343

Specifications

Chemical Name or Material 4-Nitrothiophenol
Name Note 95%
CAS 1849-36-1
CAS Min % 75.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥75%
Assay Percent Range 80%
Molecular Formula C6H5NO2S
Linear Formula SHC6H4NO
Formula Weight 155.2
MDL Number MFCD00007343
Synonym 4-Nitrobenzenethiol
Accessories

STENCH

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents