Please login to your online account to display your discounted pricing

4-Nitrothiophenol, 80%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Nitrothiophenol
Name Note 95%
CAS 1849-36-1
CAS Min % 75.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥75%
Assay Percent Range 80%
Molecular Formula C6H5NO2S
Linear Formula SHC6H4NO
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336200050

  SDS
fisher scientific acros
336200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336200250

  SDS
thermo fisher scientific
336200250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥75%
Formula Weight 155.2
Physical Form Crystalline Powder and/or Chunks
Purity Grade Technical Grade
Quantity 5g
Color Yellow
EINECS Number 217-436-4
Melting Point 72° to 80°C
RTECS Number DC1850000
Beilstein 06, 339
CAS Min % 75.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 STENCH
Packaging Glass bottle
Synonym 4-Nitrobenzenethiol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Linear Formula SHC6H4NO
Molecular Formula C6H5NO2S
Chemical Name or Material 4-Nitrothiophenol
CAS 1849-36-1
MDL Number MFCD00007343
Assay Percent Range 80%
Name Note 95%
Safety and Handling

STENCH

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.