missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrothioanisole 98.0+%, TCI America™
Supplier: TCI America N076125G
Specifications
4-Nitrothioanisole | |
71°C | |
140°C | |
25 g | |
NEZGPRYOJVPJKL-UHFFFAOYSA-N | |
1-methylsulfanyl-4-nitrobenzene | |
96109 | |
≥98.0% (GC) |
701-57-5 | |
Yellow | |
C7H7NO2S | |
4-nitrothioanisole, 1-methylthio-4-nitrobenzene, methyl 4-nitrophenyl sulfide, 1-methylsulfanyl-4-nitrobenzene, benzene, 1-methylthio-4-nitro, methyl 4-nitrophenyl sulphide, 1-methylsulfanyl-4-nitro-benzene, pubchem10631, 4-methylthio nitrobenzene | |
CSC1=CC=C(C=C1)[N+](=O)[O-] | |
169.198 | |
169.20 | |
Crystalline Powder |
Chemical Identifiers
701-57-5 | |
169.198 | |
4-nitrothioanisole, 1-methylthio-4-nitrobenzene, methyl 4-nitrophenyl sulfide, 1-methylsulfanyl-4-nitrobenzene, benzene, 1-methylthio-4-nitro, methyl 4-nitrophenyl sulphide, 1-methylsulfanyl-4-nitro-benzene, pubchem10631, 4-methylthio nitrobenzene | |
1-methylsulfanyl-4-nitrobenzene |
C7H7NO2S | |
NEZGPRYOJVPJKL-UHFFFAOYSA-N | |
96109 | |
CSC1=CC=C(C=C1)[N+](=O)[O-] |
Safety and Handling
TSCA : No