missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitroquinoline-N-oxide, 98%, ACROS Organics™

Manufacturer:  Acros Organics 203792500

 View more versions of this product

Catalog No. AC203792500



Specifications

Specifications

4-Nitroquinoline-N-oxide
Glass bottle
154°C to 156°C
56-57-5
100.0
C9H6N2O3
4 nqo
YHQDZJICGQWFHK-UHFFFAOYSA-N
4-nitro-1-oxidoquinolin-1-ium
5955
190.16
≥97.5%
Authentic
Brown to Ochre-Yellow
250mg
97.5
98%
MFCD00006738
Solubility in water: .. Other solubilities: soluble in acetone,dmso,dmf
C1=CC=C2C(=C1)C(=CC=[N+]2[O-])[N+](=O)[O-]
190.16
CHEBI:16907
Crystalline Solid

Chemical Identifiers

56-57-5
190.16
YHQDZJICGQWFHK-UHFFFAOYSA-N
5955
4-nitro-1-oxidoquinolin-1-ium
C9H6N2O3
MFCD00006738
4 nqo
CHEBI:16907
C1=CC=C2C(=C1)C(=CC=[N+]2[O-])[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS P Statement
May cause cancer.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents