Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitroquinoline N-oxide, 98+%

$49.73 - $114.66

Chemical Identifiers

Chemical Name or Material 4-Nitroquinoline N-oxide
CAS 56-57-5
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006738
Beilstein 165756
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0260503

  SDS
alfa aesar™
L0260503
1g Each for $114.66

AAL02605MD

  SDS
alfa aesar™
L02605MD
250mg Each for $49.73
Description & Specifications

Specifications

Formula Weight 190.16
Chemical Name or Material 4-Nitroquinoline N-oxide
CAS 56-57-5
Assay ≥98%
MDL Number MFCD00006738
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H340-H350
May cause genetic defects.
May cause cancer.
RTECS Number VC2100000
Beilstein 165756
Quantity 1g
Molecular Formula C9H6N2O3
Melting Point 154° to 156°C
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H340-H350
May cause genetic defects.
May cause cancer.