Please login to your online account to display your discounted pricing

4-Nitropyridine N-oxide, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128920050

 View more versions of this product

Catalog No. AC128920050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 140.1
Physical Form Crystals or Powder
Quantity 5g
Color Yellow to brown
RTECS Number UT6380000
TSCA TSCA
EINECS Number 214-395-4
Melting Point 159° to 164°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Nitropyridine N-oxide
Molecular Formula C5H4N2O3
MDL Number MFCD00006206
CAS 1124-33-0

Specifications

Chemical Name or Material 4-Nitropyridine N-oxide
CAS 1124-33-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H4N2O3
Formula Weight 140.1
MDL Number MFCD00006206
Accessories

GHS Signal Word: Warning

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents