missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitropyrazole, TCI America™

$19.86 - $42.78

Chemical Identifiers

CAS 2075-46-9
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.076
MDL Number MFCD00159626
InChI Key XORHNJQEWQGXCN-UHFFFAOYSA-N
Synonym 4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984
PubChem CID 16376
IUPAC Name 4-nitro-1H-pyrazole
SMILES C1=C(C=NN1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N08625G View Documents TCI America
N08625G
5g Each for $19.86
 
N086225G View Documents TCI America
N086225G
25g Each for $42.78
 
Specifications

Chemical Identifiers

2075-46-9
113.076
XORHNJQEWQGXCN-UHFFFAOYSA-N
16376
C1=C(C=NN1)[N+](=O)[O-]
C3H3N3O2
MFCD00159626
4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984
4-nitro-1H-pyrazole

Specifications

323°C
162°C
2075-46-9
MFCD00159626
XORHNJQEWQGXCN-UHFFFAOYSA-N
4-nitro-1H-pyrazole
16376
Crystal-Powder at 20°C
4-Nitropyrazole
White
5g
C3H3N3O2
4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984
C1=C(C=NN1)[N+](=O)[O-]
113.076
113.08
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : UQ7880000

TSCA : No