missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitropyrazole 98.0+%, TCI America™
Supplier: TCI America N086225G
Specifications
4-Nitropyrazole | |
162°C | |
323°C | |
MFCD00159626 | |
4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984 | |
[O-][N+](=O)C1=CNN=C1 | |
113.08 | |
113.08 | |
Crystalline Powder |
2075-46-9 | |
White | |
C3H3N3O2 | |
25 g | |
XORHNJQEWQGXCN-UHFFFAOYSA-N | |
4-nitro-1H-pyrazole | |
16376 | |
≥98.0% (GC,T) |
Chemical Identifiers
2075-46-9 | |
113.08 | |
XORHNJQEWQGXCN-UHFFFAOYSA-N | |
16376 | |
[O-][N+](=O)C1=CNN=C1 |
C3H3N3O2 | |
MFCD00159626 | |
4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984 | |
4-nitro-1H-pyrazole |
Safety and Handling
RTECSNumber : UQ7880000
TSCA : No