missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophthalimide 98.0+%, TCI America™
$51.43 - $497.70
Chemical Identifiers
CAS | 89-40-7 |
---|---|
Molecular Formula | C8H4N2O4 |
Molecular Weight (g/mol) | 192.13 |
MDL Number | MFCD00005884 |
InChI Key | ANYWGXDASKQYAD-UHFFFAOYSA-N |
Synonym | 4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione |
PubChem CID | 6969 |
IUPAC Name | 5-nitroisoindole-1,3-dione |
SMILES | C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O |
Chemical Identifiers
89-40-7 | |
192.13 | |
ANYWGXDASKQYAD-UHFFFAOYSA-N | |
6969 | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O |
C8H4N2O4 | |
MFCD00005884 | |
4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione | |
5-nitroisoindole-1,3-dione |
Specifications
89-40-7 | |
White-Yellow | |
MFCD00005884 | |
4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O | |
192.13 | |
192.13 | |
Crystalline Powder |
202°C | |
C8H4N2O4 | |
25 g | |
ANYWGXDASKQYAD-UHFFFAOYSA-N | |
5-nitroisoindole-1,3-dione | |
6969 | |
≥98.0% (HPLC,T) | |
4-Nitrophthalimide |
Safety and Handling
EINECSNumber : (5)-0081
RTECSNumber : TI5625000
TSCA : Yes