Please login to your online account to display your discounted pricing

4-Nitrophthalic acid, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC416125000

 View more versions of this product

Catalog No. AC416125000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (HPLC)
Color Yellow to beige
Formula Weight 211.13
Physical Form Crystalline Powder
Quantity 500g
Melting Point 163° to 167°C
Packaging Glass bottle
CAS 610-27-5
Chemical Name or Material 4-Nitrophthalic acid
Infrared Spectrum Authentic
Molecular Formula C8H5NO6
Assay Percent Range 99%

Specifications

Chemical Name or Material 4-Nitrophthalic acid
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C8H5NO6
Formula Weight 211.13