missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyloxamic acid, 98%

$57.36 - $174.26

Chemical Identifiers

CAS 103-94-6
Molecular Formula C8H6N2O5
Molecular Weight (g/mol) 210.145
MDL Number MFCD00014709
InChI Key ZEJKSYPGUAUQKW-UHFFFAOYSA-N
Synonym 4-nitrophenyloxamic acid, oxalyl-p-nitroaniline, n-4-nitrophenyl oxamic acid, 4'-nitrooxanilic acid, 2-4-nitrophenyl amino-2-oxoacetic acid, acetic acid, 4-nitrophenyl amino oxo, oxanilic acid, 4'-nitro, 4-nitrophenyl amino oxoacetic acid, 2-4-nitroanilino-2-oxoacetic acid, kyselina n-4-nitrofenyl oxamova czech
PubChem CID 7686
IUPAC Name 2-(4-nitroanilino)-2-oxoacetic acid
SMILES C1=CC(=CC=C1NC(=O)C(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0032503 View Documents Alfa Aesar™
L0032503
1g Each for $57.36
 
AAL0032506 View Documents Alfa Aesar™
L0032506
5g Each for $174.26
 
Specifications

Chemical Identifiers

103-94-6
210.145
ZEJKSYPGUAUQKW-UHFFFAOYSA-N
7686
C1=CC(=CC=C1NC(=O)C(=O)O)[N+](=O)[O-]
C8H6N2O5
MFCD00014709
4-nitrophenyloxamic acid, oxalyl-p-nitroaniline, n-4-nitrophenyl oxamic acid, 4'-nitrooxanilic acid, 2-4-nitrophenyl amino-2-oxoacetic acid, acetic acid, 4-nitrophenyl amino oxo, oxanilic acid, 4'-nitro, 4-nitrophenyl amino oxoacetic acid, 2-4-nitroanilino-2-oxoacetic acid, kyselina n-4-nitrofenyl oxamova czech
2-(4-nitroanilino)-2-oxoacetic acid

Specifications

205°C
103-94-6
MFCD00014709
4-nitrophenyloxamic acid, oxalyl-p-nitroaniline, n-4-nitrophenyl oxamic acid, 4'-nitrooxanilic acid, 2-4-nitrophenyl amino-2-oxoacetic acid, acetic acid, 4-nitrophenyl amino oxo, oxanilic acid, 4'-nitro, 4-nitrophenyl amino oxoacetic acid, 2-4-nitroanilino-2-oxoacetic acid, kyselina n-4-nitrofenyl oxamova czech
C1=CC(=CC=C1NC(=O)C(=O)O)[N+](=O)[O-]
210.145
210.15
4-Nitrophenyloxamic acid
1g
C8H6N2O5
2809447
ZEJKSYPGUAUQKW-UHFFFAOYSA-N
2-(4-nitroanilino)-2-oxoacetic acid
7686
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RP3377000

TSCA : TSCA