Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Nitrophenylketoximino)carbonyl chloride, polymer-supported, 0.8-1.0 mmol/g on polystyrene

Chemical Identifiers

Chemical Name or Material (4-Nitrophenylketoximino)carbonyl chloride
Name Note polymer-supported, 0.8 to 1.0mmol/g on polystyrene
MDL Number MFCD03791359
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1975303

 
alfa aesar™
L1975303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1975306

 
alfa aesar™
L1975306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1975309

 
alfa aesar™
L1975309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (4-Nitrophenylketoximino)carbonyl chloride
MDL Number MFCD03791359
Name Note polymer-supported, 0.8 to 1.0mmol/g on polystyrene
Quantity 1g