Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g

Chemical Identifiers

Chemical Name or Material 4-Nitrophenylketoxime on polystrene
Name Note 2% cross-linked
MDL Number MFCD00165078
Synonym Kaiser Oxime resin; 4-Nitrobenzophenone oxime, polymer supported
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1937606

 
alfa aesar™
L1937606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1937603

 
alfa aesar™
L1937603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Nitrophenylketoxime on polystrene
Synonym Kaiser Oxime resin; 4-Nitrobenzophenone oxime, polymer supported
Mesh Size 200 to 400 mesh
Name Note 2% cross-linked
MDL Number MFCD00165078
Concentration 0.8 to 1.0mmol/g
Quantity 5g