Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenylglyoxal hydrate, 97%

Chemical Identifiers

Chemical Name or Material 4-Nitrophenylglyoxal hydrate
CAS 4974-57-6
Formula Weight 179.13 (Anhydrous)
MDL Number MFCD07776822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3500903

 
alfa aesar™
H3500903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3500909

 
alfa aesar™
H3500909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.13 (Anhydrous)
Melting Point 96° to 98°C
Chemical Name or Material 4-Nitrophenylglyoxal hydrate
Physical Form Crystalline Powder
Assay 97%
Color Cream
MDL Number MFCD07776822
CAS 4974-57-6
Quantity 1g

Applications
As important industrial compounds and are mainly used as solvents. A new method for arginase assay

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.